Assay Details
Target Gene Details
Entrez Gene ID: | 73166 |
Gene Name: | transmembrane 7 superfamily member 2 |
Gene Aliases: |
3110041O18Rik, ANG1, C14SR, Dhcr14 |
Location: |
Chr.19:6062821-6067850 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tm7sf2 | NM_028454.2 | 5 | 682 | NP_082730.2 |
| AF480070.1 | 5 | 556 | AAQ05836.1 | |
| AK014166.1 | 5 | 663 | BAB29187.1 | |
| AK037325.1 | 4 | 486 | ||
| AK039028.1 | 5 | 612 | ||
| AK040016.1 | 5 | 558 | BAE20564.1 | |
| AK043687.1 | 3 | 949 | ||
| AK134512.1 | 1 | 1467 | ||
| AK155475.1 | 5 | 639 | BAE33285.1 | |
| BC014769.1 | 5 | 674 | AAH14769.1 |
More Information
Set Membership: |
|

Genomic Map