Assay Details
Target Gene Details
Entrez Gene ID: | 73166 |
Gene Name: | transmembrane 7 superfamily member 2 |
Gene Aliases: |
3110041O18Rik, ANG1, C14SR, Dhcr14 |
Location: |
Chr.19:6062821-6067850 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tm7sf2 | NM_028454.2 | NP_082730.2 | ||
| AF480070.1 | AAQ05836.1 | |||
| AI846153.1 | ||||
| AK014166.1 | BAB29187.1 | |||
| AK155475.1 | BAE33285.1 | |||
| BC014769.1 | AAH14769.1 | |||
| BY551700.1 |
More Information
Set Membership: |
|

Genomic Map