Assay Details
Target Gene Details
Entrez Gene ID: | 77836 |
Gene Name: | melan-A |
Gene Aliases: |
A930034P04Rik, Mart1 |
Location: |
Chr.19:29697548-29708447 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mlana | NM_029993.1 | NP_084269.1 | ||
| AK020928.1 | BAB32259.1 | |||
| BC111114.1 | AAI11115.1 |

Genomic Map