Assay Details
Target Gene Details
Entrez Gene ID: | 70769 |
Gene Name: | nucleolar and coiled-body phosphoprotein 1 |
Gene Aliases: |
NOPP130, NOPP140, P130 |
Location: |
Chr.19:46075847-46085543 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 7 - Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nolc1 | NM_001039351.2 | NP_001034440.1 | ||
| NM_001039352.2 | NP_001034441.1 | |||
| NM_001039353.2 | NP_001034442.1 | |||
| NM_053086.3 | NP_444316.2 | |||
| AK014336.1 | ||||
| AK029150.1 | BAC26327.1 | |||
| AK034817.1 | ||||
| AK129040.1 | BAC97850.1 | |||
| AK147118.1 | BAE27690.1 | |||
| AK150332.1 | BAE29474.1 | |||
| AK151031.1 | BAE30048.1 | |||
| AK152941.1 | BAE31612.1 | |||
| AK166753.1 | BAE38994.1 |

Genomic Map