Assay Details
Target Gene Details
Entrez Gene ID: | 225888 |
Gene Name: | suppressor of variegation 4-20 homolog 1 (Drosophila) |
Gene Aliases: |
AA117471, C630029K18Rik, KMT5B, Suv4-20h1 |
Location: |
Chr.19:3764155-3818303 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Suv420h1 | NM_001167885.1 | NP_001161357.1 | ||
| NM_001167886.1 | NP_001161358.1 | |||
| NM_001167887.1 | NP_001161359.1 | |||
| NM_001167889.1 | 11 | 1856 | NP_001161361.1 | |
| NM_144871.4 | NP_659120.3 | |||
| AK082660.1 | 10 | 1536 | BAC38565.1 | |
| AK083227.1 | BAC38817.1 | |||
| AK139255.1 | BAE23934.1 | |||
| AK152179.1 | BAE31010.1 | |||
| AY555192.1 | AAT00539.1 | |||
| BC075709.1 | AAH75709.1 |

Genomic Map