Assay Details
Target Gene Details
Entrez Gene ID: | 13409 |
Gene Name: | transmembrane channel-like gene family 1 |
Gene Aliases: |
4933416G09Rik, Beethoven, Bth, CWEA1, dn |
Location: |
Chr.19:20783458-21037152 on Build GRCm38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tmc1 | NM_028953.2 | 17 | 1956 | NP_083229.1 |
| AF417579.1 | 17 | 1956 | AAL86400.1 | |
| AK016832.1 | 3 | 307 | ||
| AK077146.1 | 3 | 316 | BAC36642.1 | |
| AY263155.1 | 3 | 230 | AAP35263.1 |

Genomic Map