Assay Details
Target Gene Details
Entrez Gene ID: | 18947 |
Gene Name: | pancreatic lipase-related protein 2 |
Gene Aliases: |
PLRP2 |
Location: |
Chr.19:58759723-58777534 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pnliprp2 | NM_011128.2 | 1 | 80 | NP_035258.2 |
| AK008021.1 | 1 | 85 | ||
| AK131882.1 | 1 | 66 | BAE20849.1 | |
| BC094923.1 | 1 | 39 | AAH94923.1 | |
| M30687.1 | 1 | 74 | AAA37491.1 |
More Information
Set Membership: |
|

Genomic Map