Assay Details
Target Gene Details
Entrez Gene ID: | 76308 |
Gene Name: | RAB1B, member RAS oncogene family |
Gene Aliases: |
1110011F09Rik |
Location: |
Chr.19:5099207-5106996 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rab1b | NM_029576.3 | NP_083852.1 | ||
| AB232584.1 | BAF02846.1 | |||
| AK003609.1 | BAB22888.1 | |||
| AK042020.1 | BAE20616.1 | |||
| AK156726.1 | BAE33821.1 | |||
| AK208578.1 | ||||
| BC016408.1 | AAH16408.1 |

Genomic Map