Assay Details
Target Gene Details
Entrez Gene ID: | 18120 |
Gene Name: | mitochondrial ribosomal protein L49 |
Gene Aliases: |
4833419D18Rik, Nof1 |
Location: |
Chr.19:6053630-6057751 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mrpl49 | NM_026246.3 | NP_080522.1 | ||
| AK003040.1 | ||||
| AK007712.1 | BAB25205.1 | |||
| AK011273.1 | BAC25326.1 | |||
| AK014723.1 | BAB29520.1 | |||
| AK027949.1 | BAC25678.1 | |||
| AK049794.1 | BAC33922.1 | |||
| AK077397.1 | BAC36782.1 | |||
| AK150572.1 | BAE29667.1 | |||
| AK150715.1 | BAE29793.1 | |||
| AK210880.1 | ||||
| AK217865.1 | ||||
| BC028956.1 | AAH28956.1 |
More Information
Set Membership: |
|

Genomic Map