Assay Details
Target Gene Details
Entrez Gene ID: | 240518 |
Gene Name: | pellino 3 |
Gene Aliases: |
6030441F14Rik, A930011L17, BC028931 |
Location: |
Chr.19:4929717-4943127 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Peli3 | NM_172835.3 | NP_766423.3 | ||
| AK044418.1 | BAC31913.1 | |||
| AK135575.1 | ||||
| BC028931.1 | AAH28931.1 |
More Information
Set Membership: |
|

Genomic Map