Assay Details
Target Gene Details
Entrez Gene ID: | 100039957 |
Gene Name: | interferon induced transmembrane protein 2 pseudogene |
Gene Aliases: |
D630022E15Rik |
Location: |
Chr.19:8774485-8776962 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm2518 | NR_015538.1 | 1 | 1002 | |
| AK041085.1 | 2 | 901 | ||
| AK077736.1 | 1 | 998 | ||
| AK082962.1 | 1 | 1001 | ||
| AK085400.1 | 1 | 974 |
Target Gene Details
Entrez Gene ID: | 225895 |
Gene Name: | TAF6-like RNA polymerase II, p300/CBP-associated factor (PCAF)-associated factor |
Gene Aliases: |
2810417N14Rik, BB223262, C530024J06Rik, Paf65a |
Location: |
Chr.19:8774354-8786417 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Taf6l | NM_001177798.1 | NP_001171269.1 | ||
| NM_146092.2 | NP_666204.2 | |||
| AK077149.1 | BAC36645.1 | |||
| BC028647.1 | AAH28647.1 |
More Information
Set Membership: |
|

Genomic Map