Assay Details
Target Gene Details
Entrez Gene ID: | 67706 |
Gene Name: | transmembrane protein 179B |
Gene Aliases: |
0610031L02Rik, 1500003K22Rik, 1810059G22Rik |
Location: |
Chr.19:8772522-8774467 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tmem179b | NM_026325.3 | NP_080601.2 | ||
| NR_028418.1 | ||||
| NR_028419.1 | ||||
| AK002731.1 | ||||
| AK005135.1 | ||||
| AK007906.1 | BAB25340.1 | |||
| AK010996.1 | BAB27315.1 | |||
| BC116961.1 | ||||
| BC116963.1 |
More Information
Set Membership: |
|

Genomic Map