Assay Details
Target Gene Details
Entrez Gene ID: | 73466 |
Gene Name: | membrane-spanning 4-domains, subfamily A, member 13 |
Gene Aliases: |
1700060E18Rik, Tetm4.2 |
Location: |
Chr.19:11169418-11196755 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ms4a13 | NM_198224.3 | 1 | 137 | NP_937867.2 |
| AF544402.1 | 1 | 142 | AAQ11744.1 | |
| AK006841.1 | 1 | 152 | ||
| BC089483.1 | 1 | 137 | AAH89483.1 |
More Information
Set Membership: |
|

Genomic Map