Assay Details
Target Gene Details
Entrez Gene ID: | 108995 |
Gene Name: | TBC1 domain family, member 10c |
Gene Aliases: |
1810062O14Rik, AI428527 |
Location: |
Chr.19:4184357-4191195 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tbc1d10c | NM_178650.3 | 3 | 256 | NP_848765.2 |
| AK040487.1 | 3 | 257 | BAC30605.1 | |
| AK075803.1 | 3 | 243 | BAC35971.1 | |
| AK131177.1 | 2 | 237 | BAD21427.1 | |
| AK146426.1 | 3 | 250 | BAE27160.1 | |
| AK153836.1 | 3 | 254 | ||
| AK154444.1 | 2 | 234 | ||
| BQ829173.1 | 3 | 492 |
More Information
Set Membership: |
|

Genomic Map