Assay Details
Target Gene Details
Entrez Gene ID: | 108995 |
Gene Name: | TBC1 domain family, member 10c |
Gene Aliases: |
1810062O14Rik, AI428527 |
Location: |
Chr.19:4184357-4191195 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tbc1d10c | NM_178650.3 | NP_848765.2 | ||
| AK040487.1 | BAC30605.1 | |||
| AK075803.1 | BAC35971.1 | |||
| AK131177.1 | BAD21427.1 | |||
| AK146426.1 | BAE27160.1 | |||
| AK153836.1 | ||||
| AK154444.1 |
More Information
Set Membership: |
|

Genomic Map