Assay Details
Target Gene Details
Entrez Gene ID: | 170813 |
Gene Name: | membrane-spanning 4-domains, subfamily A, member 3 |
Gene Aliases: |
HTm4 |
Location: |
Chr.19:11629499-11640851 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ms4a3 | NM_133246.5 | NP_573509.1 | ||
| AF280401.1 | AAK37993.1 | |||
| AF321128.1 | AAL16955.1 | |||
| AK036464.1 | BAC29440.1 | |||
| AK158372.1 | BAE34478.1 | |||
| AY184359.1 | AAN77872.1 | |||
| BC117013.1 | ||||
| BC117015.1 | ||||
| BC144853.1 |
More Information
Set Membership: |
|

Genomic Map