Assay Details
Target Gene Details
Entrez Gene ID: | 19662 |
Gene Name: | retinol binding protein 4, plasma |
Gene Aliases: |
Rbp-4 |
Location: |
Chr.19:38116620-38125321 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rbp4 | NM_001159487.1 | NP_001152959.1 | ||
| NM_011255.3 | NP_035385.1 | |||
| AK004839.1 | ||||
| AK008765.1 | BAB25881.1 | |||
| BC093529.1 | AAH93529.1 | |||
| BY112174.1 | ||||
| BY769772.1 |
More Information
Set Membership: |
|

Genomic Map