Assay Details
Target Gene Details
Entrez Gene ID: | 225997 |
Gene Name: | transient receptor potential cation channel, subfamily M, member 6 |
Gene Aliases: |
CHAK2, TRPM6/CHAK2 |
Location: |
Chr.19:18730178-18892516 on Build GRCm38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Trpm6 | NM_153417.1 | 18 | 2381 | NP_700466.1 |
| AK080899.1 | 18 | 2381 | ||
| AY135644.1 | 18 | 2381 | AAN15217.1 |
More Information
Set Membership: |
|

Genomic Map