Assay Details
Target Gene Details
Entrez Gene ID: | 225849 |
Gene Name: | protein phosphatase 2, regulatory subunit B', beta |
Gene Aliases: |
B'beta, BC026670 |
Location: |
Chr.19:6227767-6235840 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 12 - Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ppp2r5b | NM_198168.3 | NP_937811.1 | ||
| AK034167.1 | BAE20490.1 | |||
| AK155872.1 | ||||
| AK171041.1 | BAE42206.1 | |||
| AK171178.1 | BAE42296.1 | |||
| AK171430.1 | ||||
| AK196369.1 | ||||
| BC026670.1 | AAH26670.1 | |||
| BC058977.1 | AAH58977.1 |
More Information
Set Membership: |
|

Genomic Map