Assay Details
Target Gene Details
Entrez Gene ID: | 102466780 |
Gene Name: | microRNA 6986 |
Gene Aliases: |
Gm27709, mmu-mir-6986 |
Location: |
Chr.19:4623898-4623955 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mir6986 | NR_105952.1 |
Target Gene Details
Entrez Gene ID: | 19671 |
Gene Name: | RCE1 homolog, prenyl protein peptidase (S. cerevisiae) |
Gene Aliases: |
D19Ertd283e, D19Ertd98e |
Location: |
Chr.19:4622551-4625617 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rce1 | NM_023131.1 | NP_075620.1 | ||
| AJ251645.1 | CAC17014.1 | |||
| AK012946.1 | BAB28566.1 | |||
| BC004060.1 | AAH04060.1 |
More Information
Set Membership: |
|

Genomic Map