Assay Details
Target Gene Details
Entrez Gene ID: | 64380 |
Gene Name: | membrane-spanning 4-domains, subfamily A, member 4C |
Gene Aliases: |
5830413L19Rik, AI449185, Ms4a9 |
Location: |
Chr.19:11404717-11427246 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ms4a4c | NM_029499.3 | 3 | 332 | NP_083775.1 |
| AB026045.1 | 3 | 319 | BAB18757.1 | |
| AF237914.2 | 3 | 306 | AAK37596.2 | |
| AK017928.1 | 3 | 312 | BAB31007.1 | |
| AK151045.1 | 3 | 325 | BAE30062.1 | |
| AK153495.1 | 2 | 176 | BAE32042.1 | |
| AK172650.1 | 3 | 304 | BAE43112.1 | |
| BC027496.1 | 3 | 288 | AAH27496.1 | |
| CT010170.1 | 2 | 200 | CAJ18378.1 |
More Information
Set Membership: |
|

Genomic Map