Assay Details
Target Gene Details
Entrez Gene ID: | 104383 |
Gene Name: | REST corepressor 2 |
Gene Aliases: |
1A13, AW122124, CoREST, Rcor, Rcor1 |
Location: |
Chr.19:7267330-7275225 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rcor2 | NM_054048.3 | NP_473389.1 | ||
| AK052295.1 | BAC34922.1 | |||
| BC055719.1 | AAH55719.1 | |||
| X83587.2 | CAB93943.1 |

Genomic Map