Assay Details
Target Gene Details
Entrez Gene ID: | 20521 |
Gene Name: | solute carrier family 22 (organic anion/cation transporter), member 12 |
Gene Aliases: |
AI987855, OAT4L, Rst, Slc22al2, URAT1 |
Location: |
Chr.19:6535854-6543070 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc22a12 | NM_009203.3 | 9 | 1813 | NP_033229.3 |
| AB005451.1 | 9 | 1594 | BAA23875.1 | |
| AI314749.1 | 2 | 418 | ||
| AK142580.1 | 8 | 962 |
More Information
Set Membership: |
|

Genomic Map