Assay Details
Target Gene Details
Entrez Gene ID: | 76479 |
Gene Name: | survival motor neuron domain containing 1 |
Gene Aliases: |
2410004J23Rik, 4933440I19Rik |
Location: |
Chr.19:53379214-53391819 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Smndc1 | NM_172429.2 | NP_766017.1 | ||
| AK010395.1 | ||||
| AK077225.1 | ||||
| AK078834.1 | BAC37414.1 | |||
| AK090047.1 | BAC41064.1 | |||
| AK168575.1 | BAE40445.1 | |||
| BC058779.1 | AAH58779.1 | |||
| BC098223.1 | AAH98223.1 |
More Information
Set Membership: |
|

Genomic Map