Assay Details
Target Gene Details
Entrez Gene ID: | 69024 |
Gene Name: | sorting nexin 15 |
Gene Aliases: |
1500032B08Rik, E130013C21Rik |
Location: |
Chr.19:6119399-6133783 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Snx15 | NM_026912.1 | 5 | 514 | NP_081188.1 |
| BC016091.1 | 5 | 514 | AAH16091.1 |
More Information
Set Membership: |
|

Genomic Map