Assay Details
Target Gene Details
Entrez Gene ID: | 245857 |
Gene Name: | slingshot homolog 3 (Drosophila) |
Gene Aliases: |
BC028922, SSH-3, mSSH-3L |
Location: |
Chr.19:4261668-4270276 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ssh3 | NM_198113.2 | 12 | 1391 | NP_932781.1 |
| AB099289.1 | 12 | 1389 | BAC97812.1 | |
| AK038981.1 | 10 | 2605 | ||
| AK148293.1 | 12 | 1384 | ||
| AK149880.1 | 12 | 1412 | BAE29141.1 | |
| AK164662.1 | 11 | 1089 | ||
| AK166342.1 | 12 | 1398 | ||
| AK203571.1 | ||||
| BC028922.1 | 12 | 1391 | AAH28922.1 |
More Information
Set Membership: |
|

Genomic Map