Assay Details
Target Gene Details
Entrez Gene ID: | 78755 |
Gene Name: | family with sequence similarity 122, member B |
Gene Aliases: |
4632404H22Rik, AI256344, SPACIA2 |
Location: |
Chr.X:53243415-53269850 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam122b | NM_001166365.2 | NP_001159837.2 | ||
| NM_001166583.1 | NP_001160055.1 | |||
| NM_030167.4 | NP_084443.1 | |||
| AB541013.1 | ||||
| AK019480.1 | BAB31749.1 | |||
| AK036417.1 | BAC29421.1 | |||
| AK044896.1 | BAC32133.1 | |||
| BC066167.1 | AAH66167.1 |

Genomic Map