Assay Details
Target Gene Details
Entrez Gene ID: | 75096 |
Gene Name: | RIKEN cDNA 4930524O05 gene |
Gene Aliases: |
- |
Location: |
Chr.19:10645694-10648053 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 4930524O05Rik | NR_045316.1 | |||
| AK015890.1 |
Target Gene Details
Entrez Gene ID: | 71768 |
Gene Name: | von Willebrand factor C and EGF domains |
Gene Aliases: |
1300015B04Rik |
Location: |
Chr.19:10634229-10665210 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 9 - Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Vwce | NM_027913.1 | NP_082189.1 | ||
| AK005010.1 | ||||
| AK153937.1 | BAE32265.1 | |||
| BC043473.1 | AAH43473.1 |

Genomic Map