Assay Details
Target Gene Details
Entrez Gene ID: | 70392 |
Gene Name: | ankyrin repeat and SOCS box-containing 12 |
Gene Aliases: |
2310036C05Rik, Asb-12 |
Location: |
Chr.X:95470199-95557993 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Asb12 | NM_080858.3 | NP_543134.1 | ||
| AF403040.1 | AAL57359.1 | |||
| AK009644.1 | BAB26410.1 | |||
| BC107257.2 | AAI07258.1 |

Genomic Map