Assay Details
Target Gene Details
Entrez Gene ID: | 73866 |
Gene Name: | family with sequence similarity 122, member C |
Gene Aliases: |
4930432H15Rik, 4930527G05Rik |
Location: |
Chr.X:53273419-53331186 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 7 - Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam122c | NM_028671.2 | NP_082947.1 | ||
| AK015288.1 | BAB29780.1 | |||
| AK015916.1 | BAB30029.1 | |||
| BC147640.1 | ||||
| BC147646.1 | ||||
| BC172114.1 |

Genomic Map