Assay Details
Target Gene Details
Entrez Gene ID: | 11666 |
Gene Name: | ATP-binding cassette, sub-family D (ALD), member 1 |
Gene Aliases: |
ALDP, Ald, Aldgh |
Location: |
Chr.X:73716597-73738287 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Abcd1 | NM_007435.1 | NP_031461.1 | ||
| AK088792.1 | BAC40574.1 | |||
| BC011273.1 | AAH11273.1 | |||
| BC079840.1 | AAH79840.1 | |||
| Z33637.1 | CAA83917.1 |
More Information
Set Membership: |
|

Genomic Map