Assay Details
Target Gene Details
Entrez Gene ID: | 11836 |
Gene Name: | v-raf murine sarcoma 3611 viral oncogene homolog |
Gene Aliases: |
1200013E08Rik, A-Raf, AW495444, Araf1 |
Location: |
Chr.X:20797787-20860521 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 14 - Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Araf | NM_009703.2 | NP_033833.1 | ||
| AK034861.1 | ||||
| AK085823.1 | ||||
| BC004757.1 | AAH04757.1 | |||
| D00024.1 | BAA00018.1 |
More Information
Set Membership: |
|

Genomic Map