Assay Details
Target Gene Details
Entrez Gene ID: | 74168 |
Gene Name: | zinc finger, DHHC domain containing 16 |
Gene Aliases: |
1500015N03Rik, Aph2 |
Location: |
Chr.19:41933472-41944104 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zdhhc16 | NM_023740.2 | 3 | 540 | NP_076229.2 |
| AF176814.1 | 3 | 513 | AAG09269.1 | |
| AK005255.1 | 1 | 136 | ||
| AK151505.1 | 3 | 522 | BAE30456.1 | |
| AK168042.1 | 3 | 515 | BAE40024.1 | |
| BC010835.1 | 3 | 504 | AAH10835.1 | |
| CK619221.1 | 3 | 527 |
More Information
Set Membership: |
|

Genomic Map