Assay Details
Target Gene Details
Entrez Gene ID: | 74168 |
Gene Name: | zinc finger, DHHC domain containing 16 |
Gene Aliases: |
1500015N03Rik, Aph2 |
Location: |
Chr.19:41933472-41944104 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zdhhc16 | NM_023740.2 | NP_076229.2 | ||
| AF176814.1 | AAG09269.1 | |||
| AK005255.1 | ||||
| AK151505.1 | BAE30456.1 | |||
| AK168042.1 | BAE40024.1 | |||
| AK179921.1 | ||||
| AK183281.1 | ||||
| AK186864.1 | ||||
| AK186908.1 | ||||
| AK188228.1 | ||||
| AK202579.1 | ||||
| AK204700.1 | ||||
| AK205856.1 | ||||
| AK207655.1 | ||||
| AK211692.1 | ||||
| AK214551.1 | ||||
| AK218025.1 | ||||
| AK219188.1 | ||||
| BC010835.1 | AAH10835.1 |
More Information
Set Membership: |
|

Genomic Map