Assay Details
Target Gene Details
Entrez Gene ID: | 320940 |
Gene Name: | ATPase, class VI, type 11C |
Gene Aliases: |
A330005H02Rik, AI315324, Ig |
Location: |
Chr.X:60223283-60404859 on Build GRCm38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Atp11c | NM_001001798.2 | 18 | 1976 | NP_001001798.1 |
| NM_001037863.1 | 18 | 1976 | NP_001032952.1 | |
| AK039243.1 | 18 | 1977 | ||
| AK043065.1 | 18 | 1970 | ||
| AK143860.1 | 17 | 1854 | ||
| BC106087.1 | 18 | 1955 | AAI06088.1 |
More Information
Set Membership: |
|

Genomic Map