Assay Details
Target Gene Details
Entrez Gene ID: | 70237 |
Gene Name: | basic helix-loop-helix domain containing, class B9 |
Gene Aliases: |
2700087I09Rik |
Location: |
Chr.X:135841639-135891081 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bhlhb9 | NM_001098222.1 | NP_001091692.1 | ||
| NM_198161.2 | NP_937804.1 | |||
| AK143861.1 | BAE25572.1 | |||
| AK157023.1 | BAE33934.1 | |||
| AK183392.1 | ||||
| AK214741.1 | ||||
| BB863588.1 | ||||
| BY272101.1 | ||||
| CF729268.1 | ||||
| CK625652.1 |
More Information
Set Membership: |
|

Genomic Map