Assay Details
Target Gene Details
Entrez Gene ID: | 209091 |
Gene Name: | cyclin B3 |
Gene Aliases: |
- |
Location: |
Chr.X:6979652-7041635 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ccnb3 | NM_183015.3 | 10 | 3734 | NP_898836.2 |
| AJ416459.1 | 2 | 131 | CAC94916.1 | |
| AJ555464.1 | 10 | 3735 | CAD88194.1 | |
| AJ555465.1 | 9 | 1010 | CAD88195.1 | |
| BC132411.1 | 9 | 3688 | ||
| BC138355.1 | 9 | 3688 |
More Information
Set Membership: |
|

Genomic Map