Assay Details
Target Gene Details
Entrez Gene ID: | 236790 |
Gene Name: | DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 26B |
Gene Aliases: |
4930535D10Rik, 6330505F04Rik, D130066O12 |
Location: |
Chr.X:56454821-56507843 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ddx26b | NM_172779.4 | 11 | 1759 | NP_766367.1 |
| AK015976.1 | 1 | 688 | ||
| AK083942.1 | 11 | 1721 | BAC39076.1 | |
| AK144026.1 | 1 | 1603 |
More Information
Set Membership: |
|

Genomic Map