Assay Details
Target Gene Details
Entrez Gene ID: | 14199 |
Gene Name: | four and a half LIM domains 1 |
Gene Aliases: |
FHL-1, KyoT, RAM14-1, SLIM, SLIM-1 |
Location: |
Chr.X:56731720-56793346 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fhl1 | NM_001077361.1 | 1 | 148 | NP_001070829.1 |
| NM_010211.3 | NP_034341.2 | |||
| AK128904.1 | ||||
| AK158966.1 | 1 | 76 | ||
| U77039.1 | AAC02727.1 |

Genomic Map