Assay Details
Target Gene Details
Entrez Gene ID: | 320940 |
Gene Name: | ATPase, class VI, type 11C |
Gene Aliases: |
A330005H02Rik, AI315324, Ig |
Location: |
Chr.X:60223283-60404859 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Atp11c | NM_001001798.2 | 6 | 642 | NP_001001798.1 |
| NM_001037863.1 | 6 | 642 | NP_001032952.1 | |
| AK039243.1 | 6 | 643 | ||
| AK043065.1 | 6 | 636 | ||
| AK079479.1 | 6 | 627 | ||
| AK143860.1 | 6 | 570 | ||
| BC106087.1 | 6 | 621 | AAI06088.1 | |
| CB244566.1 | 2 | 168 |
More Information
Set Membership: |
|

Genomic Map