Assay Details
Target Gene Details
Entrez Gene ID: | 209091 |
Gene Name: | cyclin B3 |
Gene Aliases: |
- |
Location: |
Chr.X:6979652-7041635 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ccnb3 | NM_183015.3 | NP_898836.2 | ||
| AJ416459.1 | CAC94916.1 | |||
| AJ555464.1 | CAD88194.1 | |||
| AJ555465.1 | CAD88195.1 | |||
| BC132411.1 | ||||
| BC138355.1 |
More Information
Set Membership: |
|

Genomic Map