Assay Details
Target Gene Details
Entrez Gene ID: | 22340 |
Gene Name: | vascular endothelial growth factor B |
Gene Aliases: |
VEGF-B, Vrf |
Location: |
Chr.19:6982472-6987651 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Vegfb | NM_001185164.1 | NP_001172093.1 | ||
| NM_011697.3 | NP_035827.1 | |||
| AK148188.1 | BAE28405.1 | |||
| BC046303.1 | AAH46303.1 | |||
| U43836.1 | AAC52932.1 | |||
| U43837.1 | AAC52553.1 | |||
| U48800.1 | AAB06273.1 | |||
| U52820.1 | AAC52823.1 |
More Information
Set Membership: |
|

Genomic Map