Assay Details
Target Gene Details
Entrez Gene ID: | 210766 |
Gene Name: | BRCA1/BRCA2-containing complex, subunit 3 |
Gene Aliases: |
C6.1A |
Location: |
Chr.X:75416628-75455702 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Brcc3 | NM_001166457.1 | NP_001159929.1 | ||
| NM_001166459.1 | NP_001159931.1 | |||
| NM_145956.4 | NP_666068.1 | |||
| AK011876.1 | BAB27894.1 | |||
| AK028483.1 | ||||
| AK030736.1 | BAC27105.1 | |||
| AK038919.1 | BAC30169.1 | |||
| AK149844.1 | BAE29117.1 | |||
| BC021313.1 | AAH21313.1 | |||
| BC048179.1 | AAH48179.1 | |||
| S68022.1 | AAB29006.2 |
More Information
Set Membership: |
|

Genomic Map