Assay Details
Target Gene Details
Entrez Gene ID: | 13712 |
Gene Name: | ELK1, member of ETS oncogene family |
Gene Aliases: |
Elk-1 |
Location: |
Chr.X:20933395-20950608 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Elk1 | NM_007922.5 | 2 | 299 | NP_031948.4 |
| AK082260.1 | 2 | 299 | ||
| AK132354.1 | 2 | 292 | BAE21121.1 | |
| BC054474.1 | 2 | 283 | AAH54474.1 | |
| CV561936.1 | 1 | 245 | ||
| D26066.1 | 1 | 66 | ||
| FN434115.1 | ||||
| X87257.1 | 2 | 569 | CAA60715.1 | |
| Z36939.1 | 1 | 84 | CAA85391.1 |
More Information
Set Membership: |
|

Genomic Map