Assay Details
Target Gene Details
Entrez Gene ID: | 13712 |
Gene Name: | ELK1, member of ETS oncogene family |
Gene Aliases: |
Elk-1 |
Location: |
Chr.X:20933395-20950608 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Elk1 | NM_007922.5 | NP_031948.4 | ||
| AK132354.1 | BAE21121.1 | |||
| BC054474.1 | AAH54474.1 | |||
| X87257.1 | CAA60715.1 |
More Information
Set Membership: |
|

Genomic Map