Assay Details
Target Gene Details
Entrez Gene ID: | 26370 |
Gene Name: | centrin 2 |
Gene Aliases: |
1110034A02Rik, AI326150, Calt, caltractin |
Location: |
Chr.X:72913565-72918344 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cetn2 | NM_019405.6 | NP_062278.2 | ||
| AK004081.1 | BAB23161.1 | |||
| AK010541.1 | BAB27017.1 | |||
| AK140156.1 | BAE24259.1 | |||
| AK151033.1 | BAE30050.1 |
More Information
Set Membership: |
|

Genomic Map