Assay Details
Target Gene Details
Entrez Gene ID: | 192176 |
Gene Name: | filamin, alpha |
Gene Aliases: |
ABP-280, Dilp2, F730004A14Rik, Fln1, GENA 379, filamin-1 |
Location: |
Chr.X:74223461-74249854 on Build GRCm38 |
Assay Gene Location: | Within Exon 36 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Flna | NM_001290421.1 | 36 | 5997 | NP_001277350.1 |
| NM_010227.3 | 35 | 5975 | NP_034357.2 | |
| AK131179.1 | 23 | 3816 | BAD21429.1 | |
| BC151024.1 | 35 | 5966 |

Genomic Map