Assay Details
Target Gene Details
Entrez Gene ID: | 192176 |
Gene Name: | filamin, alpha |
Gene Aliases: |
ABP-280, Dilp2, F730004A14Rik, Fln1, GENA 379, filamin-1 |
Location: |
Chr.X:74223461-74249854 on Build GRCm38 |
Assay Gene Location: | Within Exon 45 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Flna | NM_001290421.1 | 45 | 7468 | NP_001277350.1 |
| NM_010227.3 | 44 | 7446 | NP_034357.2 | |
| AK131179.1 | 32 | 5287 | BAD21429.1 | |
| BC004061.1 | 7 | 967 | AAH04061.1 | |
| BC054432.1 | 1 | 46 | AAH54432.1 | |
| BC099506.1 | 2 | 218 | AAH99506.1 | |
| BC151024.1 | 44 | 7437 |

Genomic Map