Assay Details
Target Gene Details
Entrez Gene ID: | 631784 |
Gene Name: | predicted gene 7073 |
Gene Aliases: |
- |
Location: |
Chr.X:60436052-60456195 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm7073 | NM_001039240.3 | NP_001034329.2 | ||
| AK135660.1 | BAE22603.1 |

Genomic Map