Assay Details
Target Gene Details
Entrez Gene ID: | 26926 |
Gene Name: | apoptosis-inducing factor, mitochondrion-associated 1 |
Gene Aliases: |
AIF, AIFsh2, Hq, Pdcd8 |
Location: |
Chr.X:48474944-48513563 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Aifm1 | NM_001290364.1 | NP_001277293.1 | ||
| NM_012019.3 | NP_036149.1 | |||
| AF100927.1 | AAD16435.1 | |||
| AK133764.1 | ||||
| AK211196.1 | ||||
| BC003292.1 | AAH03292.1 | |||
| BY250260.1 | ||||
| DQ016499.1 |
More Information
Set Membership: |
|

Genomic Map